siRNA Details
Gene Symbol: COL6A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278298.1 | NP_001265227.1 | 7 | 3076 | ||
NM_153264.6 | NP_694996.5 | 7 | 3076 | |||
NR_022012.2 | 7 | 3076 | ||||
XM_011512621.2 | XP_011510923.1 | 7 | 2950 | |||
XM_011512622.2 | XP_011510924.1 | 7 | 2950 | |||
XM_011512623.2 | XP_011510925.1 | 7 | 2950 | |||
GenBank mRNA | AM906079.1 | - | 4 | 1543 | ||
EU085556.1 | - | 7 | 3076 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
