siRNA Details
Gene Symbol: FOXK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037165.1 | NP_001032242.1 | 8 | 1736 | ||
XM_011515191.2 | XP_011513493.1 | 10 | 1407 | |||
GenBank mRNA | AK122663.1 | - | 9 | 1226 | ||
AK302243.1 | - | 8 | 1375 | |||
BC038434.1 | - | 11 | 1480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
