siRNA Details
Gene Symbol: FOXK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037165.1 | NP_001032242.1 | 2 | 623 | ||
XM_011515191.2 | XP_011513493.1 | 4 | 294 | |||
GenBank mRNA | AW205747.1 | - | 2 | 333 | ||
BC038434.1 | - | 5 | 367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
