siRNA Details
Gene Symbol: JRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077527.2 | NP_001070995.2 | 3 | 3372 | ||
NM_001279352.1 | NP_001266281.1 | 4 | 2631 | |||
NM_003724.3 | NP_003715.3 | 2 | 8905 | |||
XM_006716677.3 | XP_006716740.1 | 2 | 9486 | |||
XM_006716678.3 | XP_006716741.1 | 2 | 10007 | |||
GenBank mRNA | BC036794.1 | - | 2 | 4103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
