siRNA Details
Gene Symbol: JRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077527.2 | NP_001070995.2 | 3 | 2635 | ||
NM_003724.3 | NP_003715.3 | 2 | 8168 | |||
XM_006716677.3 | XP_006716740.1 | 2 | 8749 | |||
XM_006716678.3 | XP_006716741.1 | 2 | 9270 | |||
GenBank mRNA | AF072467.1 | - | 2 | 4216 | ||
AF072468.1 | - | 2 | 1691 | |||
AK024975.1 | - | 1 | 2069 | |||
AK025687.1 | - | 1 | 5728 | |||
BC036794.1 | - | 2 | 3368 | |||
BX640755.1 | - | 3 | 2635 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
