siRNA Details
Gene Symbol: OR6B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC137561.1 | - | 1 | 1007 | 9021561 | |
BC137562.1 | - | 1 | 1007 | 9021562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
