siRNA Details
Gene Symbol: SPNS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB441165.1 | - | 8, 9 | 1267 | ||
BC041772.1 | - | 8, 9 | 1480 | 4814561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
