siRNA Details
Gene Symbol: FMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103184.3 | NP_001096654.1 | 15 | 3484 | ||
NM_001277313.1 | NP_001264242.1 | 19 | 4509 | |||
XM_011521504.2 | XP_011519806.1 | 19 | 4332 | |||
XM_011521506.2 | XP_011519808.1 | 17 | 4246 | |||
XM_011521509.2 | XP_011519811.1 | 18 | 2540 | |||
XM_011521511.2 | XP_011519813.1 | 16 | 4011 | |||
XM_017022130.1 | XP_016877619.1 | 16 | 4703 | |||
XM_017022132.1 | XP_016877621.1 | 16 | 4457 | |||
XM_017022133.1 | XP_016877622.1 | 18 | 4362 | |||
XM_017022134.1 | XP_016877623.1 | 16 | 2913 | |||
XM_017022135.1 | XP_016877624.1 | 16 | 4182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
