siRNA Details
Gene Symbol: CCDC137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199287.2 | NP_954981.1 | 6 | 1444 | ||
XM_011524738.2 | XP_011523040.1 | 6 | 1346 | |||
GenBank mRNA | BC009369.2 | - | 6 | 1428 | 4110481 | |
FJ649645.1 | - | 6 | 3134 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
