siRNA Details
Gene Symbol: PTCHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034842.3 | NP_001030014.2 | 2, 3 | 1321 | ||
GenBank mRNA | BC107139.2 | - | 2, 3 | 1245 | 40026539 | |
JF332167.1 | - | 2, 3 | 1203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
