siRNA Details
Gene Symbol: DEFT1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036686.1 | 2 | 226 | |||
GenBank mRNA | AF355799.1 | - | 2 | 226 | ||
BC069283.1 | - | 1 | 129 |
Gene Symbol: DEFT1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036687.1 | 2 | 226 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
