siRNA Details
Gene Symbol: WASIR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130735.1 | 2 | 368 | |||
GenBank mRNA | AL539368.3 | - | 2 | 383 | ||
BC032901.1 | - | 2 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
