siRNA Details
Gene Symbol: MYO7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080527.1 | NP_001073996.1 | 12 | 1387 | ||
XM_006712539.3 | XP_006712602.1 | 12 | 1500 | |||
XM_011511218.2 | XP_011509520.1 | 12 | 1500 | |||
XM_017004169.1 | XP_016859658.1 | 12 | 1500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
