siRNA Details
Gene Symbol: RAB12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025300.2 | NP_001020471.2 | 6 | 1836 | ||
XM_006722300.3 | XP_006722363.1 | 6 | 2149 | |||
GenBank mRNA | AK127824.1 | - | 5 | 1451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
