siRNA Details
Gene Symbol: KIAA2026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017969.2 | NP_001017969.2 | 8 | 4227 | ||
XM_005251385.2 | XP_005251442.1 | 7 | 4613 | |||
XM_006716730.3 | XP_006716793.1 | 7 | 2251 | |||
XM_011517759.2 | XP_011516061.1 | 8 | 4703 | |||
XM_011517760.2 | XP_011516062.1 | 8 | 4703 | |||
XM_011517761.2 | XP_011516063.1 | 5 | 2073 | |||
XM_017014331.1 | XP_016869820.1 | 8 | 4703 | |||
XM_017014332.1 | XP_016869821.1 | 4 | 2176 | |||
XM_017014333.1 | XP_016869822.1 | 5 | 1825 | |||
XM_017014334.1 | XP_016869823.1 | 4 | 1932 | |||
GenBank mRNA | AB095946.1 | - | 1 | 276 | ||
BC136846.1 | - | 3 | 1558 | 9020837 | ||
BX648372.1 | - | 4 | 1710 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
