siRNA Details
Gene Symbol: CYLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271680.1 | NP_001258609.1 | 3 | 207 | ||
NM_021118.2 | NP_066941.1 | 3 | 210 | |||
XM_005262086.3 | XP_005262143.1 | 3 | 207 | |||
GenBank mRNA | BC021669.1 | - | 3 | 194 | ||
BC058019.1 | - | 3 | 194 | |||
BC070355.1 | - | 3 | 187 | |||
BC126461.1 | - | 3 | 174 | 8992177 | ||
BX110674.1 | - | 3 | 168 | |||
DB028115.1 | - | 3 | 207 | |||
DB049397.1 | - | 3 | 210 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
