siRNA Details
Gene Symbol: CYLC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021118.2 | NP_066941.1 | 4 | 358 | ||
XM_005262086.3 | XP_005262143.1 | 4 | 355 | |||
GenBank mRNA | BC021669.1 | - | 4 | 342 | ||
BC038787.1 | - | 1 | 20 | |||
BC058019.1 | - | 4 | 342 | |||
BC070355.1 | - | 4 | 335 | |||
BC126461.1 | - | 4 | 322 | 8992177 | ||
DB028115.1 | - | 4 | 355 | |||
DB049397.1 | - | 4 | 358 | |||
Z22780.1 | - | 2 | 143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
