siRNA Details
Gene Symbol: TRIM61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012414.2 | NP_001012414.1 | 3 | 904 | ||
GenBank mRNA | AK296100.1 | - | 1 | 777 | ||
BC089393.1 | - | 3 | 904 | 30378540 |
Gene Symbol: FARSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003110.1 | XP_016858599.1 | 3 | 425 | ||
GenBank mRNA | AK001025.1 | - | 3 | 414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
