siRNA Details
Gene Symbol: CIITA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720880.3 | XP_006720943.2 | 1 | 255 | ||
XM_011522484.2 | XP_011520786.1 | 1 | 255 | |||
XM_011522485.2 | XP_011520787.1 | 1 | 255 | |||
XM_011522486.2 | XP_011520788.1 | 1 | 257 | |||
XM_011522487.2 | XP_011520789.1 | 1 | 139 | |||
XM_011522489.2 | XP_011520791.1 | 1 | 139 | |||
XM_011522491.2 | XP_011520793.1 | 1 | 257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
