siRNA Details
Gene Symbol: C10orf120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010912.2 | NP_001010912.1 | 3 | 809 | ||
GenBank mRNA | BC065903.1 | - | 3 | 813 | ||
BC140922.1 | - | 3 | 801 | 9056734 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
