siRNA Details
Gene Symbol: KIF26B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018012.3 | NP_060482.2 | 3 | 1338 | ||
GenBank mRNA | AK125187.1 | - | 3 | 1339 | ||
AK293964.1 | - | 3 | 1253 | |||
AK297303.1 | - | 3 | 1334 | |||
AY923834.1 | - | 3 | 1339 | |||
BC042481.1 | - | 2 | 672 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
