siRNA Details
Gene Symbol: C6orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010905.2 | NP_001010905.1 | 1 | 249 | ||
GenBank mRNA | AK303850.1 | - | 1 | 249 | ||
BC062712.1 | - | 1 | 253 | 30353154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
