siRNA Details
Gene Symbol: DCAF8L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136533.1 | NP_001130005.1 | 1 | 338 | ||
GenBank mRNA | BC157859.1 | - | 1 | 338 | 9057078 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
