siRNA Details
Gene Symbol: EXOC3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126522.1 | 1 | 348 | |||
GenBank mRNA | BC014011.2 | - | 1 | 283 | ||
BC029796.1 | - | 1 | 331 | 5183143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
