siRNA Details
Gene Symbol: ZKSCAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012981.4 | NP_001012999.3 | 1 | 793 | ||
XM_017023200.1 | XP_016878689.1 | 1 | 1202 | |||
GenBank mRNA | AK131342.1 | - | 1 | 793 | ||
AK302399.1 | - | 1 | 818 | |||
AK303472.1 | - | 1 | 1190 | |||
BC130007.1 | - | 1 | 451 | 40132986 | ||
BC151139.1 | - | 1 | 437 | 9057039 | ||
BX648785.1 | - | 1 | 781 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
