siRNA Details
Gene Symbol: THEGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256475.1 | NP_001243404.1 | 6 | 1025 | ||
XM_011534357.1 | XP_011532659.1 | 6 | 1059 | |||
XM_011534361.2 | XP_011532663.1 | 6 | 1059 | |||
XM_011534363.2 | XP_011532665.1 | 6 | 1059 | |||
XM_011534364.1 | XP_011532666.1 | 6 | 1059 | |||
XM_011534365.2 | XP_011532667.1 | 6 | 1059 | |||
GenBank mRNA | DY655427.1 | - | 1 | 17 | ||
DY655539.1 | - | 2 | 248 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
