siRNA Details
Gene Symbol: KLHL38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081675.2 | NP_001075144.2 | 1 | 293 | ||
XM_005250901.4 | XP_005250958.1 | 2 | 514 | |||
GenBank mRNA | AB196636.1 | - | 1 | 269 | ||
BC127886.1 | - | 1 | 293 | 40131758 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
