siRNA Details
Gene Symbol: GATSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037666.2 | NP_001032755.1 | 9 | 1295 | ||
GenBank mRNA | AK055587.1 | - | 9 | 1201 | ||
BC021927.1 | - | 9 | 1181 | 4330223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
