siRNA Details
Gene Symbol: GATSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037666.2 | NP_001032755.1 | 3 | 479 | ||
GenBank mRNA | AK055587.1 | - | 3 | 385 | ||
AK309436.1 | - | 2 | 312 | |||
AK310246.1 | - | 2 | 327 | |||
BC021927.1 | - | 3 | 365 | 4330223 | ||
CR456449.1 | - | 3 | 394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
