siRNA Details
Gene Symbol: FMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305424.1 | NP_001292353.1 | 16 | 5124 | ||
NM_020066.4 | NP_064450.3 | 15 | 5112 | |||
XM_017001839.1 | XP_016857328.1 | 16 | 3964 | |||
XM_017001840.1 | XP_016857329.1 | 15 | 3364 | |||
XM_017001841.1 | XP_016857330.1 | 15 | 3822 | |||
XM_017001842.1 | XP_016857331.1 | 14 | 3043 | |||
XM_017001843.1 | XP_016857332.1 | 13 | 3031 | |||
GenBank mRNA | AB209153.1 | - | 16 | 3717 | ||
AF218942.1 | - | 7 | 612 | |||
AF225426.1 | - | 7 | 634 | |||
AK297716.1 | - | 10 | 882 | |||
AK298141.1 | - | 8 | 947 | |||
BC014364.2 | - | 5 | 451 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
