siRNA Details
Gene Symbol: YIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039671.2 | NP_001034760.1 | 8 | 1518 | ||
NM_001039672.2 | NP_001034761.1 | 8 | 1536 | |||
NM_001039673.2 | NP_001034762.1 | 8 | 1527 | |||
NM_001145461.1 | NP_001138933.1 | 8 | 1512 | |||
NM_001145462.1 | NP_001138934.1 | 8 | 1488 | |||
NM_001145463.1 | NP_001138935.1 | 9 | 1604 | |||
GenBank mRNA | AK027130.1 | - | 1 | 211 | ||
BC025257.1 | - | 1 | 147 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
