siRNA Details
Gene Symbol: TENM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080477.3 | NP_001073946.1 | 2 | 260 | ||
XM_017008385.1 | XP_016863874.1 | 5 | 698 | |||
XM_017008386.1 | XP_016863875.1 | 2 | 285 | |||
XM_017008387.1 | XP_016863876.1 | 2 | 258 | |||
XM_017008388.1 | XP_016863877.1 | 2 | 233 | |||
XM_017008389.1 | XP_016863878.1 | 5 | 698 | |||
XM_017008390.1 | XP_016863879.1 | 5 | 698 | |||
XM_017008391.1 | XP_016863880.1 | 5 | 698 | |||
XM_017008392.1 | XP_016863881.1 | 5 | 698 | |||
XM_017008393.1 | XP_016863882.1 | 5 | 698 | |||
XM_017008394.1 | XP_016863883.1 | 5 | 698 | |||
GenBank mRNA | AF195420.1 | - | 2 | 289 | ||
AK308489.1 | - | 2 | 389 | |||
HE578284.1 | - | 1 | 139 | |||
HE578285.1 | - | 1 | 139 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
