siRNA Details
Gene Symbol: VMA21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017980.3 | NP_001017980.1 | 3 | 2166 | ||
XM_011531125.2 | XP_011529427.1 | 3 | 2492 | |||
GenBank mRNA | AK091301.1 | - | 1 | 874 | ||
AL833596.1 | - | 3 | 2119 | |||
BC110800.1 | - | 3 | 2079 | 6044374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
