siRNA Details
Gene Symbol: C12orf71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080406.1 | NP_001073875.1 | 1 | 415 | ||
XM_011520847.2 | XP_011519149.1 | 2 | 957 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
