siRNA Details
Gene Symbol: CYP2B7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001278.1 | 2, 3 | 343 | |||
GenBank mRNA | AK307933.1 | - | 2, 3 | 389 | ||
AK310944.1 | - | 2, 3 | 393 | |||
DQ198366.1 | - | 2, 3 | 335 | |||
DQ198367.1 | - | 2, 3 | 335 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
