siRNA Details
Gene Symbol: CCDC85C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144995.1 | NP_001138467.1 | 6 | 5101 | ||
GenBank mRNA | AK026211.1 | - | 1 | 732 | ||
BC031990.1 | - | 1 | 501 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
