siRNA Details
Gene Symbol: MGAT4EP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038135.2 | 3 | 1975 | |||
GenBank mRNA | BC032956.1 | - | 3 | 1972 | 4827085 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
