siRNA Details
Gene Symbol: TMEM198B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036476.1 | 5 | 2379 | |||
NR_036477.1 | 4 | 2286 | ||||
NR_036478.1 | 6 | 2035 | ||||
NR_036479.1 | 5 | 1891 | ||||
GenBank mRNA | AF370398.1 | - | 2 | 1057 | ||
AK057179.1 | - | 4 | 1207 | |||
AK094261.1 | - | 1 | 1060 | |||
AK095340.1 | - | 5 | 1754 | |||
BC072431.1 | - | 5 | 923 | |||
BX648969.1 | - | 3 | 1128 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
