siRNA Details
Gene Symbol: TOM1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033551.2 | NP_001028723.1 | 14 | 5320 | ||
NM_001082968.1 | NP_001076437.1 | 15 | 5470 | |||
NM_001288786.1 | NP_001275715.1 | 15 | 5398 | |||
NM_001288787.1 | NP_001275716.1 | 14 | 5335 | |||
NM_001288788.1 | NP_001275717.1 | 12 | 5116 | |||
NM_001288789.1 | NP_001275718.1 | 13 | 5113 | |||
XM_005256461.2 | XP_005256518.1 | 16 | 5491 | |||
XM_005256462.1 | XP_005256519.1 | 15 | 5358 | |||
XM_005256463.2 | XP_005256520.1 | 14 | 5344 | |||
XM_005256466.1 | XP_005256523.1 | 14 | 5199 | |||
XM_011523662.1 | XP_011521964.1 | 15 | 5343 | |||
XM_017024201.1 | XP_016879690.1 | 13 | 5211 | |||
GenBank mRNA | AK055959.1 | - | 1 | 3215 | ||
AL133641.1 | - | 1 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
