siRNA Details
Gene Symbol: TOM1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033551.2 | NP_001028723.1 | 14 | 5140 | ||
NM_001082968.1 | NP_001076437.1 | 15 | 5290 | |||
NM_001288786.1 | NP_001275715.1 | 15 | 5218 | |||
NM_001288787.1 | NP_001275716.1 | 14 | 5155 | |||
NM_001288788.1 | NP_001275717.1 | 12 | 4936 | |||
NM_001288789.1 | NP_001275718.1 | 13 | 4933 | |||
XM_005256461.2 | XP_005256518.1 | 16 | 5311 | |||
XM_005256462.1 | XP_005256519.1 | 15 | 5178 | |||
XM_005256463.2 | XP_005256520.1 | 14 | 5164 | |||
XM_005256466.1 | XP_005256523.1 | 14 | 5019 | |||
XM_011523662.1 | XP_011521964.1 | 15 | 5163 | |||
XM_017024201.1 | XP_016879690.1 | 13 | 5031 | |||
GenBank mRNA | AK055959.1 | - | 1 | 3035 | ||
AL133641.1 | - | 1 | 196 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
