siRNA Details
Gene Symbol: LINC00889 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026935.1 | 1 | 775 | |||
GenBank mRNA | AK055234.1 | - | 1 | 324 | ||
AK055382.1 | - | 1 | 459 | |||
BC105719.1 | - | 1 | 449 | |||
BC105725.1 | - | 1 | 449 | |||
BC105726.1 | - | 1 | 449 | |||
BC105727.1 | - | 1 | 449 | |||
BC139831.1 | - | 1 | 452 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
