siRNA Details
Gene Symbol: ARFGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020340.4 | NP_065073.3 | 34 | 13608 | ||
GenBank mRNA | AB252196.1 | - | 34 | 13608 | ||
BC022980.1 | - | 1 | 2126 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
