siRNA Details
Gene Symbol: MYADML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145113.2 | NP_001138585.2 | 3 | 1355 | ||
GenBank mRNA | BC029306.1 | - | 3 | 1355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
