siRNA Details
Gene Symbol: ZNF829 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037232.3 | NP_001032309.2 | 6 | 762 | ||
NM_001171979.1 | NP_001165450.1 | 6 | 707 | |||
XM_005258876.4 | XP_005258933.1 | 6 | 585 | |||
XM_011526933.2 | XP_011525235.1 | 6 | 604 | |||
GenBank mRNA | AK130969.1 | - | 5 | 442 | ||
BC107131.1 | - | 6 | 649 | 40025539 | ||
BC107132.1 | - | 6 | 653 | 40025541 | ||
BX538288.1 | - | 6 | 708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
