siRNA Details
Gene Symbol: LRRC69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129890.1 | NP_001123362.1 | 7 | 936 | ||
GenBank mRNA | AK097393.1 | - | 10 | 1311 | ||
AK130865.1 | - | 3 | 442 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
