siRNA Details
Gene Symbol: HIGD1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109619.1 | NP_001103089.1 | 2 | 213 | ||
XM_011538649.2 | XP_011536951.1 | 4 | 360 | |||
XM_017019783.1 | XP_016875272.1 | 5 | 1788 | |||
XM_017019784.1 | XP_016875273.1 | 5 | 1788 | |||
XM_017019785.1 | XP_016875274.1 | 3 | 292 | |||
GenBank mRNA | DW009649.1 | - | 2 | 150 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
