siRNA Details
Gene Symbol: HIGD1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109619.1 | NP_001103089.1 | 2, 3 | 225 | ||
XM_017019784.1 | XP_016875273.1 | 5, 6 | 1800 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
