siRNA Details
Gene Symbol: EYS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142800.1 | NP_001136272.1 | 24 | 4194 | ||
NM_001292009.1 | NP_001278938.1 | 24 | 4194 | |||
GenBank mRNA | FJ416331.1 | - | 24 | 4202 | ||
FM209056.1 | - | 24 | 4146 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
