siRNA Details
Gene Symbol: SDR42E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017023977.1 | XP_016879466.1 | 9 | 1781 | ||
XM_017023978.1 | XP_016879467.1 | 8 | 2542 | |||
XM_017023979.1 | XP_016879468.1 | 8 | 2542 | |||
XM_017023980.1 | XP_016879469.1 | 8 | 2542 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
