siRNA Details
Gene Symbol: SMIM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289970.1 | NP_001276899.1 | 4 | 304 | ||
XM_011534622.1 | XP_011532924.1 | 4 | 387 | |||
XM_011534623.2 | XP_011532925.1 | 4 | 498 | |||
XM_011534624.2 | XP_011532926.1 | 5 | 386 | |||
GenBank mRNA | EG327678.1 | - | 4 | 249 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
